Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nortropine 98.0+%, TCI America™
SDP

Catalog No. N08435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08435G 5 g
N084325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08435G Supplier TCI America Supplier No. N08435G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 538-09-0
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.19
MDL Number MFCD00047140,MFCD01711246,MFCD21642368
InChI Key YYMCYJLIYNNOMK-UHFFFAOYNA-N
Synonym 8-Azabicyclo[3.2.1]octan-3-ol
PubChem CID 73480
IUPAC Name 8-azabicyclo[3.2.1]octan-3-ol
SMILES OC1CC2CCC(C1)N2

Specifications

Quantity 5 g
Formula Weight 127.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Nortropine
RTECSNumber RD0700000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.