Learn More
Cayman Chemical NorursodeoxycholIc AcId 10mg
SDP
Catalog No. 502946841
Encompass_Preferred
A bile acid; is secreted in an unchanged form, as trihydroxy derivatives, or as sulfate or glucuronide conjugates; decreases serum levels of ALT and alkaline phosphatase, periductal fibrosis, and hepatic hydroxyproline content in an Mdr2-/- mouse model of sclerosing cholangitis at 0.5% w/w in the diet
Catalog No. 50-294-6841 Supplier Cayman Chemical Supplier No. 3392510 MG
A bile acid; is secreted in an unchanged form, as trihydroxy derivatives, or as sulfate or glucuronide conjugates; decreases serum levels of ALT and alkaline phosphatase, periductal fibrosis, and hepatic hydroxyproline content in an Mdr2-/- mouse model of sclerosing cholangitis at 0.5% w/w in the diet