Learn More
Cayman Chemical noruscogenIn 10mg
SDP
Catalog No. 502971632
Encompass_Preferred
A natural sapogenin that acts as a bioavailable, potent, and high-affinity agonist of the nuclear receptor RORα (EC50 = 110 nM); up-regulates the expression of several RORα-inducible genes in the livers of mice, when given at 3 mg/kg/d orally for seven days
Catalog No. 50-297-1632 Supplier Cayman Chemical Supplier No. 1556710 MG
A natural sapogenin that acts as a bioavailable, potent, and high-affinity agonist of the nuclear receptor RORα (EC50 = 110 nM); up-regulates the expression of several RORα-inducible genes in the livers of mice, when given at 3 mg/kg/d orally for seven days