Learn More
Medchemexpress LLC Norvancomycin (monohydrochloride) | 213997-73-0 | 95.3% | 1 ML
SDP
Catalog No. 5000412187
Encompass_Preferred
Norvancomycin monohydrochloride is a cell wall synthesis inhibitor, targeting peptidoglycan precursors of Gram-positive bacteria. It irreversibly inhibits bacterial cell wall synthesis, showing antibacterial activity. Primarily used for Gram-positive bacterial infections like MRSA and MRSE, it can also be incorporated into bionic calcium phosphate coatings on titanium implants to enhance antibacterial activity and prevent postoperative orthopedic infections.
  • Targets peptidoglycan precursors of Gram-positive bacteria
  • Cannot pass the blood-brain barrier
  • Inhibits bacterial cell wall synthesis
  • Effective against Gram-positive bacterial infections (MRSA, MRSE)
  • Enhances antibacterial activity in titanium implant coatings
  • Inhibits Staphylococcus aureus in in vitro studies
  • Widely distributed in rat tissues (not in brain)
Catalog No. 50-004-12187 Supplier Medchemexpress LLC Supplier No. HYB192410MM/1ML
Norvancomycin monohydrochloride is a cell wall synthesis inhibitor, targeting peptidoglycan precursors of Gram-positive bacteria. It irreversibly inhibits bacterial cell wall synthesis, showing antibacterial activity. Primarily used for Gram-positive bacterial infections like MRSA and MRSE, it can also be incorporated into bionic calcium phosphate coatings on titanium implants to enhance antibacterial activity and prevent postoperative orthopedic infections.
  • Targets peptidoglycan precursors of Gram-positive bacteria
  • Cannot pass the blood-brain barrier
  • Inhibits bacterial cell wall synthesis
  • Effective against Gram-positive bacterial infections (MRSA, MRSE)
  • Enhances antibacterial activity in titanium implant coatings
  • Inhibits Staphylococcus aureus in in vitro studies
  • Widely distributed in rat tissues (not in brain)