Learn More
Medchemexpress LLC Norvancomycin monohydrochloride | 213997-73-0 | 95.3% | 1471.69 g/mol | C65H74Cl3N9O24 | 10 MG
SDP
Catalog No. 5000295537
Encompass_Preferred
Norvancomycin monohydrochloride is a glycopeptide antibiotic (N-demethylvancomycin) supplied as a hydrochloride salt for research use. It competitively inhibits bacterial cell wall synthesis in Gram-positive organisms and is provided in milligram-scale quantities for in vitro studies and material-coating applications.
  • High-purity research reagent with reported purity of ~95.3%.
  • Provided as a hydrochloride salt in powder form for easy dissolution.
  • Used to study Gram-positive bacteria, including methicillin-resistant strains.
  • Available in multiple milligram pack sizes suitable for laboratory experiments.
  • Documentation available: datasheet, safety data sheet, and certificate of analysis.
Catalog No. 50-002-95537 Supplier Medchemexpress LLC Supplier No. HYN13150A10MG
Norvancomycin monohydrochloride is a glycopeptide antibiotic (N-demethylvancomycin) supplied as a hydrochloride salt for research use. It competitively inhibits bacterial cell wall synthesis in Gram-positive organisms and is provided in milligram-scale quantities for in vitro studies and material-coating applications.
  • High-purity research reagent with reported purity of ~95.3%.
  • Provided as a hydrochloride salt in powder form for easy dissolution.
  • Used to study Gram-positive bacteria, including methicillin-resistant strains.
  • Available in multiple milligram pack sizes suitable for laboratory experiments.
  • Documentation available: datasheet, safety data sheet, and certificate of analysis.