Learn More
Medchemexpress LLC Norverapamil-d7 hydrochloride | 1216413-74-9 | 98.8% | 484.08 g/mol | C26H30D7ClN2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY135328AS5MG
Encompass_Preferred
Norverapamil-d7 hydrochloride is a deuterium-labeled analogue of norverapamil used as an analytical tracer and internal standard for quantitative assays. It is intended for research applications involving L-type calcium channels and P-glycoprotein function and is compatible with common mass spectrometry and NMR workflows.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity suitable for analytical workflows.
  • Solid form for convenient handling.
  • Stable storage: -20°C sealed; in solvent: -80°C (6 months), -20°C (1 month).
  • Can be used as a tracer or internal standard for NMR, GC-MS, and LC-MS.
Catalog No. 50-002-22631