Learn More
Medchemexpress LLC Norverapamil (hydrochloride) | 67812-42-4 | MFCD00078605 | 99.8% | 477.04 g·mol⁻¹ | C26H37ClN2O4 | 1 MG
SDP
Catalog No. 5000213156
Encompass_Preferred
Norverapamil hydrochloride is the monohydrochloride salt of an N-demethylated metabolite of verapamil. It acts as an L-type calcium channel blocker and an inhibitor of P-glycoprotein (P-gp), and is used in pharmacological and transporter research.
  • Acts as an L-type calcium channel blocker.
  • Inhibits P-glycoprotein (P-gp) function.
  • Suitable for in vitro pharmacology and transporter studies.
  • Supplied as a solid or as a DMSO solution for experimental use.
  • High reported purity (99.8%).
  • Molecular weight 477.04 g·mol⁻¹; formula C26H37ClN2O4.
Catalog No. 50-002-13156 Supplier Medchemexpress LLC Supplier No. HY1007501MG
Norverapamil hydrochloride is the monohydrochloride salt of an N-demethylated metabolite of verapamil. It acts as an L-type calcium channel blocker and an inhibitor of P-glycoprotein (P-gp), and is used in pharmacological and transporter research.
  • Acts as an L-type calcium channel blocker.
  • Inhibits P-glycoprotein (P-gp) function.
  • Suitable for in vitro pharmacology and transporter studies.
  • Supplied as a solid or as a DMSO solution for experimental use.
  • High reported purity (99.8%).
  • Molecular weight 477.04 g·mol⁻¹; formula C26H37ClN2O4.