Learn More
Medchemexpress LLC Norverapamil (hydrochloride) | 67812-42-4 | MFCD00078605 | 99.8% | 477.04 g/mol | C26H37ClN2O4 | 1 ML

Supplier: Medchemexpress LLC HY10075010MM 1ML
Norverapamil hydrochloride is the N-demethylated major metabolite of verapamil that functions as an L-type calcium channel blocker and a P-glycoprotein (P-gp) inhibitor. It is provided for research use and is supplied as a white to off-white solid or as a pre-made 10 mM solution in DMSO for laboratory assays and in vivo studies.
- High purity suitable for research applications.
- Available as solid and as a 10 mM stock solution in DMSO.
- Soluble in water and DMSO; stock preparation examples provided.
- Storage recommendations for solids and solutions to preserve stability.
- Useful for calcium channel pharmacology and P-gp transporter studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.