Learn More
Medchemexpress LLC Norverapamil hydrochloride | 67812-42-4 | MFCD00078605 | 99.8% | 477.04 g/mol | C26H37ClN2O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100750100MG
Encompass_Preferred
Norverapamil hydrochloride is the N-demethylated metabolite of verapamil. It functions as an L-type calcium channel blocker and a P-glycoprotein (P-gp) function inhibitor and is supplied for research use in solid form or as a DMSO solution.
  • High purity: 99.8% for reliable experimental results.
  • White to off-white solid for easy handling.
  • Soluble in water (≥50 mg/mL) and DMSO (≥31 mg/mL) for formulation flexibility.
  • Stable when stored sealed at 4°C; in solvent store -80°C (6 months) or -20°C (1 month).
  • Suitable as a research reagent for studies of calcium channel modulation and P-gp function.
Catalog No. 50-002-11366