Learn More
Medchemexpress LLC Norverapamil hydrochloride | 67812-42-4 | MFCD00078605 | 99.8% | 477.04 g/mol | C26H37ClN2O4 | 5 MG
SDP
Catalog No. 5000212189
Encompass_Preferred
Norverapamil hydrochloride is the N-demethylated metabolite of verapamil used in pharmacological research. It functions as an L-type calcium channel blocker and a P-glycoprotein (P-gp) function inhibitor, supplied as a research-grade chemical for in vitro and in vivo studies.
  • L-type calcium channel blocker and P-gp inhibitor.
  • High purity (99.8%).
  • Molecular formula C26H37ClN2O4; molecular weight 477.04 g/mol.
  • Suitable for pharmacological and membrane transport studies.
  • Provided as a solid hydrochloride salt for straightforward handling and dosing.
Catalog No. 50-002-12189 Supplier Medchemexpress LLC Supplier No. HY1007505MG
Norverapamil hydrochloride is the N-demethylated metabolite of verapamil used in pharmacological research. It functions as an L-type calcium channel blocker and a P-glycoprotein (P-gp) function inhibitor, supplied as a research-grade chemical for in vitro and in vivo studies.
  • L-type calcium channel blocker and P-gp inhibitor.
  • High purity (99.8%).
  • Molecular formula C26H37ClN2O4; molecular weight 477.04 g/mol.
  • Suitable for pharmacological and membrane transport studies.
  • Provided as a solid hydrochloride salt for straightforward handling and dosing.