Learn More
Cayman Chemical NOS ActIvIty asy KIt 1ea
SDP
Catalog No. 502897036
Encompass_Preferred
The NOS Activity Assay measures NOS activity by monitoring the conversion of radiolabeled arginine to citrulline. This assay is simple, sensitive, and specific for NOS activity and can be used with both crude and purified enzyme preparations. The kit includes sufficient materials and reagents for 50 total reactions. Radiolabeled arginine and NADPH are not included with the kit.
Catalog No. 50-289-7036 Supplier Cayman Chemical Supplier No. 7810011 EA
Add to Cart
Add to Cart
The NOS Activity Assay measures NOS activity by monitoring the conversion of radiolabeled arginine to citrulline. This assay is simple, sensitive, and specific for NOS activity and can be used with both crude and purified enzyme preparations. The kit includes sufficient materials and reagents for 50 total reactions. Radiolabeled arginine and NADPH are not included with the kit.