Learn More
Cayman Chemical nNOS Electrophoresis Standard, Purity: ≥95%, 1 ea
SDP
Catalog No. 502912834
Encompass_Preferred
Each vial contains nNOS (rat) to be used as a standard for WB and electrophoresis. The enzyme has been carefully purified to exclude all unrelated proteins; may not be catalytically active.{1281}
Catalog No. 50-291-2834 Supplier Cayman Chemical Supplier No. 3608701 EA
Each vial contains nNOS (rat) to be used as a standard for WB and electrophoresis. The enzyme has been carefully purified to exclude all unrelated proteins; may not be catalytically active.{1281}