Learn More
Enzo Life Sciences Nitric oxide synthase (inducible) polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA2000100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 1131-1141 of mouse macrophage NOS. Species reactivity: Human, Mouse, Rat. Applications: ICC, IHC, WB. Formulation: Liquid. Neat serum.
Catalog No. 50-201-1767