Learn More
Cayman Chemical nOS InhIbItr I 1mg
SDP
Catalog No. 502983131
Encompass_Preferred
A cell-permeable, selective inhibitor of nNOS (Ki = 120 nM)
Catalog No. 50-298-3131 Supplier Cayman Chemical Supplier No. 209061 MG
Add to Cart
Add to Cart
A cell-permeable, selective inhibitor of nNOS (Ki = 120 nM)