Learn More
Cayman Chemical nOS InhIbItr I 500ug
SDP
Catalog No. 502981696
Encompass_Preferred
A cell-permeable, selective inhibitor of nNOS (Ki = 120 nM)
Catalog No. 50-298-1696 Supplier Cayman Chemical Supplier No. 20906500 UG
A cell-permeable, selective inhibitor of nNOS (Ki = 120 nM)