Learn More
Enzo Life Sciences Nitric oxide synthase (universal) monoclonal antibody (NOS-3F7-B11-B5) (200µl)
SDP
Supplier:  Enzo Life Sciences ALX804029R200
Encompass_Preferred
Clone: NOS-3F7-B11-B5. Isotype: IgM. Immunogen: Purified neuronal nitric oxide synthase (nNOS; NOS I) from bovine brain. Species reactivity: Mouse, Rat, Bovine. Specificity: Recognizes iNOS (NOS II), eNOS (NOS III) and nNOS (NOS I). Applications: IHC (FS), WB. Formulation: Liquid. Ascites diluted in PBS containing 0.05% sodium azide.
Catalog No. 50-200-9755