Learn More
Enzo Life Sciences Nitric oxide synthase (neuronal) polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ALX210529C100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 1411-1425 (CN1411RLRSESIAFIEESK1425) of human neuronal nitric oxide synthase (nNOS; NOS I). This sequence is completely conserved in human, mouse, Rat, Rabbit and frog nNOS. UniProt ID: P29475. Species reactivity: Human, Mouse, Rat. Applications: ICC, IF, IHC, WB. Recommended Dilutions: Immunocytochemistry (1:50-1:500) Immunofluorescence (1:50-1:500) Immunohistochemistry (1:20-1:200) Western blot (1:100-1:1000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Epitope affinity-purified. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8127