Learn More
Aviva Systems Biology Corp NOS1AP Antibody - C-terminal region (ARP64061_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64061_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NOS1AP. It was validated on Western Blot. . Alternate names: 6330408P19Rik, CAPONApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-288-159