Learn More
Aviva Systems Biology Corp NOS2 Antibody - N-terminal region (ARP63000_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63000_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NOS2. It was validated on Western Blot. . Alternate names: HEP-NOS, INOS, NOS, NOS2AApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-287-838