Learn More
Brookfield Engineer Labs NOSC4-18 THERMOSEL/SSA SPINDLE
Catalog No. NC9650513
Encompass
Spindle, For use with the Thermosel System, Selection of spindal is determined by the viscosity measurement range that is required
Catalog No. NC9650513 Supplier Brookfield Engineer Labs Supplier No. SC418
Spindle, For use with the Thermosel System, Selection of spindal is determined by the viscosity measurement range that is required