Learn More
Cayman Chemical NoscapIn-13C-d3 1mg
SDP
Supplier:  Cayman Chemical 266791 MG
Encompass_Preferred
An internal standard for the quantification of noscapine by GC- or LC-MS
Catalog No. 50-293-6228