Learn More
Medchemexpress LLC Noscapine | 128-62-1 | MFCD00069316 | 413.42 | 250 MG
SDP
Catalog No. 5000349393
Encompass_Preferred
Noscapine is an orally active phthalideisoquinoline alkaloid with potent antitussive effects. It works by activating sigma opioid receptors and is a non-competitive Bradykinin inhibitor. Noscapine disrupts microtubule dynamics, induces mitotic arrest and apoptosis.
  • Possesses anticancer, neuroprotective, and anti-inflammatory activities
  • Can cross the blood-brain barrier
  • Intended for research and analytical applications
Catalog No. 50-003-49393 Supplier Medchemexpress LLC Supplier No. 250MGHY13716
Noscapine is an orally active phthalideisoquinoline alkaloid with potent antitussive effects. It works by activating sigma opioid receptors and is a non-competitive Bradykinin inhibitor. Noscapine disrupts microtubule dynamics, induces mitotic arrest and apoptosis.
  • Possesses anticancer, neuroprotective, and anti-inflammatory activities
  • Can cross the blood-brain barrier
  • Intended for research and analytical applications