Learn More
Medchemexpress LLC Noscapine (hydrochloride) | 912-60-7 | 99.9% | 449.88 | 1 G
SDP
Catalog No. 5000351721
Encompass_Preferred
Noscapine (hydrochloride) is an orally active phthalideisoquinoline alkaloid with potent antitussive properties. It activates sigma opioid receptors and acts as a non-competitive Bradykinin inhibitor. This compound also disrupts microtubule dynamics, induces mitotic arrest and apoptosis.
  • Orally active phthalideisoquinoline alkaloid
  • Potent antitussive properties
  • Activates sigma opioid receptors and acts as a non-competitive Bradykinin inhibitor
  • Disrupts microtubule dynamics, induces mitotic arrest and apoptosis
  • Exhibits anticancer, neuroprotective, and anti-inflammatory activities
  • Can cross the blood-brain barrier
Catalog No. 50-003-51721 Supplier Medchemexpress LLC Supplier No. HY13716A1G
Noscapine (hydrochloride) is an orally active phthalideisoquinoline alkaloid with potent antitussive properties. It activates sigma opioid receptors and acts as a non-competitive Bradykinin inhibitor. This compound also disrupts microtubule dynamics, induces mitotic arrest and apoptosis.
  • Orally active phthalideisoquinoline alkaloid
  • Potent antitussive properties
  • Activates sigma opioid receptors and acts as a non-competitive Bradykinin inhibitor
  • Disrupts microtubule dynamics, induces mitotic arrest and apoptosis
  • Exhibits anticancer, neuroprotective, and anti-inflammatory activities
  • Can cross the blood-brain barrier