Learn More
Medchemexpress LLC Noscapine (hydrochloride) | 912-60-7 | 99.9% | 449.88 | 250 MG
SDP
Catalog No. 5000352286
Encompass_Preferred
Noscapine hydrochloride is an orally active phthalideisoquinoline alkaloid with potent antitussive effects. It activates sigma opioid receptors and inhibits Bradykinin non-competitively. It also disrupts microtubule dynamics, leading to mitotic arrest and apoptosis. This compound exhibits anticancer, neuroprotective, and anti-inflammatory activities, and can cross the blood-brain barrier.
  • Activates sigma opioid receptors
  • Non-competitive Bradykinin inhibitor
  • Disrupts microtubule dynamics
  • Induces mitotic arrest and apoptosis
  • Exhibits anticancer activity
  • Exhibits neuroprotective activity
  • Exhibits anti-inflammatory activities
  • Capable of crossing the blood-brain barrier
Catalog No. 50-003-52286 Supplier Medchemexpress LLC Supplier No. 250MGHY13716A
Noscapine hydrochloride is an orally active phthalideisoquinoline alkaloid with potent antitussive effects. It activates sigma opioid receptors and inhibits Bradykinin non-competitively. It also disrupts microtubule dynamics, leading to mitotic arrest and apoptosis. This compound exhibits anticancer, neuroprotective, and anti-inflammatory activities, and can cross the blood-brain barrier.
  • Activates sigma opioid receptors
  • Non-competitive Bradykinin inhibitor
  • Disrupts microtubule dynamics
  • Induces mitotic arrest and apoptosis
  • Exhibits anticancer activity
  • Exhibits neuroprotective activity
  • Exhibits anti-inflammatory activities
  • Capable of crossing the blood-brain barrier