Learn More
Cayman Chemical Noscapine (hydrochloride), Purity: ≥98%, CAS: 912-60-7, 5 g
SDP
Catalog No. 502965464
Encompass_Preferred
An alkaloid with anticancer and antitussive activities; binds to tubulin and induces cell cycle arrest at the G2/M phase in HeLa cells at 20 µM; induces apoptosis in HeLa cells and thymocytes (IC50s = 25 and 10 µM, respectively) and reduces proliferation of MCF-7 breast and Renal 1983 bladder cancer cells (IC50s = 42.3 and 39.1 µM, respectively); induces tumor regression in an MCF-7 mouse xenograft model at 120 mg/kg; decreases the number of citric acid-induced coughs in guinea pigs at 30 and 70 mg/kg e
Catalog No. 50-296-5464 Supplier Cayman Chemical Supplier No. 172555 G
An alkaloid with anticancer and antitussive activities; binds to tubulin and induces cell cycle arrest at the G2/M phase in HeLa cells at 20 µM; induces apoptosis in HeLa cells and thymocytes (IC50s = 25 and 10 µM, respectively) and reduces proliferation of MCF-7 breast and Renal 1983 bladder cancer cells (IC50s = 42.3 and 39.1 µM, respectively); induces tumor regression in an MCF-7 mouse xenograft model at 120 mg/kg; decreases the number of citric acid-induced coughs in guinea pigs at 30 and 70 mg/kg e