Learn More
Hans Rudolph Inc Reusable Nose Clip
SDP
Catalog No. NC1015368
Encompass_Preferred
Nose Clips, Reusable. Reusable have a metal torsion spring and soft pad. Clips are used on the outer nose to occlude and seal off the nose breathing. One size fits adults through large pediatric subjects for use during exercise testing and respiratory research.
Catalog No. NC1015368 Supplier Hans Rudolph Inc Supplier No. 112571
Nose Clips, Reusable. Reusable have a metal torsion spring and soft pad. Clips are used on the outer nose to occlude and seal off the nose breathing. One size fits adults through large pediatric subjects for use during exercise testing and respiratory research.