Learn More
Vetequip Inc NOSE CONE 14MM RAT
SDP
Catalog No. NC9918058
Encompass_Preferred
This item has a minimum qty of 5 per supplier requirements.
Cone; Nose; Rodent-Rat; 14mm; Approximately 1-1/2in. L; Funnel-shaped cone that makes an excellent anesthesia mask by cutting down on in-mask dead space without restricting access to the eyes or cranium
Catalog No. NC9918058 Supplier Vetequip Inc Supplier No. 921614
This item has a minimum qty of 5 per supplier requirements.
Cone; Nose; Rodent-Rat; 14mm; Approximately 1-1/2in. L; Funnel-shaped cone that makes an excellent anesthesia mask by cutting down on in-mask dead space without restricting access to the eyes or cranium