Learn More
LICORbio Nose Cone Plugs for in vivo Imaging (pack of 6 large)
Catalog No. NC2225452
Encompass_Preferred
Nose cone plugs are used to block anesthesia ports that are not being used for in vivo imaging. They are available in packs of six in either small plugs designed for mouse nose cones or large plugs designed for rat nose cones.
This item is not returnable. View return policy
Catalog No. NC2225452 Supplier LICORbio Supplier No. 9000804
Nose cone plugs are used to block anesthesia ports that are not being used for in vivo imaging. They are available in packs of six in either small plugs designed for mouse nose cones or large plugs designed for rat nose cones.