Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nosiheptide | 56377-79-8 | MFCD30718117 | 97.1% | 1222.36 g/mol | C51H43N13O12S6 | 50 MG
SDP

Catalog No. 5000239724
Encompass_Preferred

Nosiheptide is a thiopeptide antibiotic that inhibits bacterial protein synthesis and is used as a research reagent and analytical standard. It is applied in studies of ribosomal function, antibiotic mechanism of action, and bacterial resistance research.

  • Thiopeptide antibiotic that inhibits bacterial protein synthesis.
  • Suitable for research and analytical applications.
  • Purity 97.05%.
  • Pack size 50 mg.
  • Molecular formula C51H43N13O12S6; molecular weight 1222.36 g/mol.
  • CAS number 56377-79-8.

Catalog No. 50-002-39724 Supplier Medchemexpress LLC Supplier No. HY10748650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nosiheptide is a thiopeptide antibiotic that inhibits bacterial protein synthesis and is used as a research reagent and analytical standard. It is applied in studies of ribosomal function, antibiotic mechanism of action, and bacterial resistance research.

  • Thiopeptide antibiotic that inhibits bacterial protein synthesis.
  • Suitable for research and analytical applications.
  • Purity 97.05%.
  • Pack size 50 mg.
  • Molecular formula C51H43N13O12S6; molecular weight 1222.36 g/mol.
  • CAS number 56377-79-8.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.