Learn More
Medchemexpress LLC HY-107486 5mg Medchemexpress, Nosiheptide CAS:56377-79-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1074865MG
Encompass_Preferred
Medchemexpress, HY-107486 5mg Nosiheptide CAS:56377-79-8 Nosiheptide (Multhiomycin), a thiopeptide antibiotic produced by Streptomyces actuosus, inhibits bacterial protein synthesis and bears a unique indole side ring system and regiospecific hydroxyl groups on the characteristic macrocyclic core. Nosiheptide has been widely used as a feed additive for animal growth. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8424