Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nosiheptide (Multhiomycin) | 56377-79-8 | MFCD00867479 | 97.1% | 1222.36 g·mol⁻¹ | C51H43N13O12S6 | 25 MG
SDP

Catalog No. 5000239721
Encompass_Preferred

Nosiheptide is a thiopeptide antibiotic produced by Streptomyces actuosus that inhibits bacterial protein synthesis. It is supplied as a research-grade standard for antibiotic studies and for investigating growth-promoting effects in animal-feed research.

  • Thiopeptide antibiotic with a distinctive indole side ring.
  • Inhibits bacterial protein synthesis by targeting elongation factors.
  • Commonly used in antimicrobial research and animal feed additive studies.
  • Supplied as a solid, light yellow to yellow powder.
  • Purity approximately 97.1%.
  • Molecular weight 1222.36 g·mol⁻¹; molecular formula C51H43N13O12S6.
  • Available in small-scale quantities suitable for laboratory use.

Catalog No. 50-002-39721 Supplier Medchemexpress LLC Supplier No. HY10748625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nosiheptide is a thiopeptide antibiotic produced by Streptomyces actuosus that inhibits bacterial protein synthesis. It is supplied as a research-grade standard for antibiotic studies and for investigating growth-promoting effects in animal-feed research.

  • Thiopeptide antibiotic with a distinctive indole side ring.
  • Inhibits bacterial protein synthesis by targeting elongation factors.
  • Commonly used in antimicrobial research and animal feed additive studies.
  • Supplied as a solid, light yellow to yellow powder.
  • Purity approximately 97.1%.
  • Molecular weight 1222.36 g·mol⁻¹; molecular formula C51H43N13O12S6.
  • Available in small-scale quantities suitable for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.