Learn More
Aviva Systems Biology Corp NOSIP Antibody - middle region (ARP56785_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56785_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NOSIP. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human NOSIP. Alternate names: CGI-25ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-214