Learn More
Med Vet International Nosorb, Non-Absorbent, Cat Litter, 5Lb
Catalog No. 502828731
Encompass_Preferred
Urinalysis is an important diagnostic tool that is sometimes overlooked in cats because of the difficulty in obtaining the sample. With Nosorb it is so easy that many veterinarians report doubling the number of urinalyses done. The indications are many, including urinary tract disease, monitoring hydration and kidney function, helping to control diabetes, part of a routine work-up, etc. Just clean the litter pan and add Nosorb. The cat will urinate as always. Nonabsorbent, inert, disposable granules make it easy for clients or technicians to obtain a urine sample Inexpensive to use and has a 98% acceptability by cats as litter Designed to be used in the hospital or to be dispensed for out-patient use.
Catalog No. 50-282-8731 Supplier Med Vet International Supplier No. PHNOSORB5LB
Urinalysis is an important diagnostic tool that is sometimes overlooked in cats because of the difficulty in obtaining the sample. With Nosorb it is so easy that many veterinarians report doubling the number of urinalyses done. The indications are many, including urinary tract disease, monitoring hydration and kidney function, helping to control diabetes, part of a routine work-up, etc. Just clean the litter pan and add Nosorb. The cat will urinate as always. Nonabsorbent, inert, disposable granules make it easy for clients or technicians to obtain a urine sample Inexpensive to use and has a 98% acceptability by cats as litter Designed to be used in the hospital or to be dispensed for out-patient use.