Learn More
Cayman Chemical nospIramycIn I 10mg
SDP
Catalog No. 502947501
Encompass_Preferred
A macrolide antibiotic; active against the macrolide-sensitive KB210 strain of S. aureus (MIC = 3.12 UG/ml), as well as B. cereus, B. subtilis, M. luteus, E. coli, and K. pneumoniae (MICs = 1.56, 3.12, 3.12, 0.2, 50, and 12.5 UG/ml, respectively; binds to E. coli ribosomes (IC50 = 1.2 µM); protects against mortality in a mouse model of S. pneumoniae type III infection (ED50 = 399.8 mg/kg)
Catalog No. 50-294-7501 Supplier Cayman Chemical Supplier No. 3406010 MG
A macrolide antibiotic; active against the macrolide-sensitive KB210 strain of S. aureus (MIC = 3.12 UG/ml), as well as B. cereus, B. subtilis, M. luteus, E. coli, and K. pneumoniae (MICs = 1.56, 3.12, 3.12, 0.2, 50, and 12.5 UG/ml, respectively; binds to E. coli ribosomes (IC50 = 1.2 µM); protects against mortality in a mouse model of S. pneumoniae type III infection (ED50 = 399.8 mg/kg)