Learn More
Cayman Chemical Neostigmine (methyl sulfate), Purity: ≥98%, CAS: 51-60-5, 1 g
SDP
Catalog No. 502994687
Encompass_Preferred
A reversible AChE inhibitor (Kd = 260 μM); dose-dependently increases the latency of paw withdrawal in response to thermal and mechanical stimuli (ED50s = 6.6 and 3.5 µg, respectively); restores muscle action potentials in mice with a thymopoietin-induced neuromuscular block (5 μg, i.p.)
Catalog No. 50-299-4687 Supplier Cayman Chemical Supplier No. 235011 G
A reversible AChE inhibitor (Kd = 260 μM); dose-dependently increases the latency of paw withdrawal in response to thermal and mechanical stimuli (ED50s = 6.6 and 3.5 µg, respectively); restores muscle action potentials in mice with a thymopoietin-induced neuromuscular block (5 μg, i.p.)