Learn More
ABclonal Technology NOSTRIN Rabbit pAb
SDP
Catalog No. 5000107743
Encompass_Preferred
Nitric oxide (NO) is a potent mediator in biologic processes such as neurotransmission, inflammatory response, and vascular homeostasis. NOSTRIN binds the enzyme responsible for NO production, endothelial NO synthase (ENOS, MIM 163729), and triggers the translocation of ENOS from the plasma membrane to vesicle-like subcellular structures, thereby attenuating ENOS-dependent NO production.
Catalog No. 50-001-07743 Supplier ABclonal Technology Supplier No. A15938500UL
Nitric oxide (NO) is a potent mediator in biologic processes such as neurotransmission, inflammatory response, and vascular homeostasis. NOSTRIN binds the enzyme responsible for NO production, endothelial NO synthase (ENOS, MIM 163729), and triggers the translocation of ENOS from the plasma membrane to vesicle-like subcellular structures, thereby attenuating ENOS-dependent NO production.