Learn More
Grainger Not Applicable 1/2 W White
Catalog No. 502749038
Encompass_Preferred
The Grainger Approved 1/2 Watt White LED Light Bulb is a long-lasting LED light bulb that produces a bright white light. It has an estimated life of 50,000 hours and consumes just 0.5 watts of power. This 120 volt LED bulb produces 25 lumens of light output. It has a candelabra screw base and measures 2.17 inches in length by 1.18 inches in diameter. This LED light bulb is ideal for accent lighting, displays, and other low wattage applications where a small white light is needed. It is cULus listed for damp locations and is manufactured in the USA.
Catalog No. 50-274-9038 Supplier Grainger Supplier No. 14A767
The Grainger Approved 1/2 Watt White LED Light Bulb is a long-lasting LED light bulb that produces a bright white light. It has an estimated life of 50,000 hours and consumes just 0.5 watts of power. This 120 volt LED bulb produces 25 lumens of light output. It has a candelabra screw base and measures 2.17 inches in length by 1.18 inches in diameter. This LED light bulb is ideal for accent lighting, displays, and other low wattage applications where a small white light is needed. It is cULus listed for damp locations and is manufactured in the USA.