Learn More
Medchemexpress LLC Not found on product page (SDS link not accessible) | 2607138-82-7 | MFCD34602688; MFCD33402167 | 99.9% | 461.79 | C19H16ClF4N3O4 | 5 MG
SDP
Catalog No. 5000221027
Encompass_Preferred
ART812 is a small-molecule, orally active inhibitor of DNA polymerase theta (Polθ) with high biochemical potency (Polθ IC50 ~7.6 nM). It exhibits cellular activity in microhomology-mediated end joining (MMEJ) assays (IC50 ~240 nM) and has been investigated preclinically for targeting homologous recombination-deficient tumors and for overcoming PARP inhibitor resistance.
  • High biochemical potency: Polθ IC50 ~7.6 nM
  • Cellular activity in MMEJ assays (IC50 ~240 nM)
  • Orally active small molecule suitable for in vivo studies
  • Mechanism targets Polθ-mediated DNA repair pathways
  • Investigated for synthetic-lethal targeting of HR-deficient tumors
  • High reported purity (~99.9%) for research use
Catalog No. 50-002-21027 Supplier Medchemexpress LLC Supplier No. HY1392895MG
ART812 is a small-molecule, orally active inhibitor of DNA polymerase theta (Polθ) with high biochemical potency (Polθ IC50 ~7.6 nM). It exhibits cellular activity in microhomology-mediated end joining (MMEJ) assays (IC50 ~240 nM) and has been investigated preclinically for targeting homologous recombination-deficient tumors and for overcoming PARP inhibitor resistance.
  • High biochemical potency: Polθ IC50 ~7.6 nM
  • Cellular activity in MMEJ assays (IC50 ~240 nM)
  • Orally active small molecule suitable for in vivo studies
  • Mechanism targets Polθ-mediated DNA repair pathways
  • Investigated for synthetic-lethal targeting of HR-deficient tumors
  • High reported purity (~99.9%) for research use