Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NOT Receptor Modulator 1 | 1015231-98-7 | 362.85 | 50 MG
SDP

Catalog No. 5000446809
Encompass_Preferred

NOT Receptor Modulator 1 is a nuclear receptor Nurr-1/NOT modulator. It was extracted from patent WO 2008034974 A1, Example 39 in table1.

  • Purity: 98.61%
  • Molecular weight: 362.85
  • Chemical formula: C22H19ClN2O
  • Appearance: White to off-white solid
  • CAS number: 1015231-98-7
  • Storage conditions: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility (in vitro): DMSO: 125 mg/mL (344.49 mM)
  • Target: Nuclear receptor NOT (EC50 = 218 nM in CHO cells)

Catalog No. 50-004-46809 Supplier Medchemexpress LLC Supplier No. HYU0042950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NOT Receptor Modulator 1 is a nuclear receptor Nurr-1/NOT modulator. It was extracted from patent WO 2008034974 A1, Example 39 in table1.

  • Purity: 98.61%
  • Molecular weight: 362.85
  • Chemical formula: C22H19ClN2O
  • Appearance: White to off-white solid
  • CAS number: 1015231-98-7
  • Storage conditions: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility (in vitro): DMSO: 125 mg/mL (344.49 mM)
  • Target: Nuclear receptor NOT (EC50 = 218 nM in CHO cells)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.