Learn More
TARGETMOL CHEMICALS INC NOT Receptor Modulator 1 5MG
SDP
Catalog No. 502876157
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.NOT Receptor Modulator 1 (2-(3-(2-(4-chlorophenyl)imidazo[1,2-a]pyridin-6-yl)phenyl)propan-2-ol) is a modulator of nuclear receptor NOT. Purity 97.01%
Catalog No. 50-287-6157 Supplier TARGETMOL CHEMICALS INC Supplier No. T122475MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.NOT Receptor Modulator 1 (2-(3-(2-(4-chlorophenyl)imidazo[1,2-a]pyridin-6-yl)phenyl)propan-2-ol) is a modulator of nuclear receptor NOT. Purity 97.01%