Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NOTA-bis(t-Butyl ester) | 1161415-28-6 | MFCD32706549 | 98.0% | 415.52 g/mol | C20H37N3O6 | 100 MG
SDP

Catalog No. 5000281231
Encompass_Preferred

NOTA-bis(tBu)ester is a NOTA-derived bifunctional chelator provided as a solid powder for research use. It is designed for bioconjugation and radiochemistry workflows where a protected NOTA scaffold is required to link ligands or drugs to metal radionuclides; the tert-butyl ester groups protect carboxylates during synthesis and can be removed in subsequent deprotection steps.

  • Bifunctional chelator suitable for drug coupling and targeted radiochemistry.
  • White to light yellow solid with reported high purity.
  • Soluble in DMSO (approx. 50 mg/mL) with ultrasonic assistance.
  • Storage stability: powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Includes a single CAS identifier for unambiguous chemical reference.

Catalog No. 50-002-81231 Supplier Medchemexpress LLC Supplier No. HYW250574100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NOTA-bis(tBu)ester is a NOTA-derived bifunctional chelator provided as a solid powder for research use. It is designed for bioconjugation and radiochemistry workflows where a protected NOTA scaffold is required to link ligands or drugs to metal radionuclides; the tert-butyl ester groups protect carboxylates during synthesis and can be removed in subsequent deprotection steps.

  • Bifunctional chelator suitable for drug coupling and targeted radiochemistry.
  • White to light yellow solid with reported high purity.
  • Soluble in DMSO (approx. 50 mg/mL) with ultrasonic assistance.
  • Storage stability: powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Includes a single CAS identifier for unambiguous chemical reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.