Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NOTA-bis(tBu)ester | 1161415-28-6 | MFCD32706549 | 98.0% | 415.52 | C20H37N3O6 | 10 MG
SDP

Catalog No. 5000281237
Encompass_Preferred

NOTA-bis(tBu)ester is a tert-butyl-protected derivative of NOTA that functions as a bifunctional chelator for radiolabeling and bioconjugation. The tert-butyl ester groups improve synthetic handling and allow conjugation steps prior to deprotection and metal chelation, supporting use in PET radiochemistry and drug conjugation workflows.

  • CAS number 1161415-28-6.
  • Molecular formula C20H37N3O6; molecular weight 415.52.
  • Purity 98.0%.
  • White powder physical form.
  • Storage: powder -20°C (long term), 4°C (short term).
  • Designed for radiolabeling and bioconjugation applications.
  • Tert-butyl protection for synthetic stability and facile deprotection.

Catalog No. 50-002-81237 Supplier Medchemexpress LLC Supplier No. HYW25057410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NOTA-bis(tBu)ester is a tert-butyl-protected derivative of NOTA that functions as a bifunctional chelator for radiolabeling and bioconjugation. The tert-butyl ester groups improve synthetic handling and allow conjugation steps prior to deprotection and metal chelation, supporting use in PET radiochemistry and drug conjugation workflows.

  • CAS number 1161415-28-6.
  • Molecular formula C20H37N3O6; molecular weight 415.52.
  • Purity 98.0%.
  • White powder physical form.
  • Storage: powder -20°C (long term), 4°C (short term).
  • Designed for radiolabeling and bioconjugation applications.
  • Tert-butyl protection for synthetic stability and facile deprotection.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.