Learn More
Medchemexpress LLC NOTA-NHS ester | 1338231-09-6 | MFCD30747764 | 90.0% | 400.38 | C16H24N4O8 | 10 MG
SDP
Catalog No. 5000209372
Encompass_Preferred
NOTA-NHS ester is an N-hydroxysuccinimide (NHS) ester used to conjugate a NOTA chelator to biomolecules for subsequent fluorescent or radioactive nuclide labeling in research applications. It is supplied as a solid reagent with defined molecular characteristics for use in radiopharmaceutical and probe development.
  • Provides an NHS-ester functional group for amine conjugation.
  • Suitable for fluorescent labeling and radioactive nuclide labeling applications.
  • Documented molecular formula C16H24N4O8 and molecular weight 400.38.
  • Available in small research pack sizes for synthesis and probe preparation.
  • Typically soluble in DMSO for conjugation reactions.
Catalog No. 50-002-09372 Supplier Medchemexpress LLC Supplier No. HYW08702810MG
NOTA-NHS ester is an N-hydroxysuccinimide (NHS) ester used to conjugate a NOTA chelator to biomolecules for subsequent fluorescent or radioactive nuclide labeling in research applications. It is supplied as a solid reagent with defined molecular characteristics for use in radiopharmaceutical and probe development.
  • Provides an NHS-ester functional group for amine conjugation.
  • Suitable for fluorescent labeling and radioactive nuclide labeling applications.
  • Documented molecular formula C16H24N4O8 and molecular weight 400.38.
  • Available in small research pack sizes for synthesis and probe preparation.
  • Typically soluble in DMSO for conjugation reactions.