Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NOTA-NHS ester | 1338231-09-6 | MFCD30747764 | ≥97.0% | 400.38 g/mol | C16H24N4O8 | 25 MG
SDP

Catalog No. 5000208870
Encompass_Preferred

NOTA-NHS ester is a bifunctional chelator derivative used for fluorescent labeling and radiolabeling (radionuclide-drug conjugates). It contains an activated N-hydroxysuccinimide (NHS) ester for conjugation to primary amines and a NOTA chelator for radiometal coordination. Supplied for research use only.

  • Activated N-hydroxysuccinimide (NHS) ester for conjugation to primary amines.
  • NOTA chelator for stable binding of radiometals.
  • Purity ≥97.0%.
  • CAS number 1338231-09-6.
  • Molecular formula C16H24N4O8; molecular weight 400.38 g/mol.
  • Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-08870 Supplier Medchemexpress LLC Supplier No. 25MGHYW087028
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NOTA-NHS ester is a bifunctional chelator derivative used for fluorescent labeling and radiolabeling (radionuclide-drug conjugates). It contains an activated N-hydroxysuccinimide (NHS) ester for conjugation to primary amines and a NOTA chelator for radiometal coordination. Supplied for research use only.

  • Activated N-hydroxysuccinimide (NHS) ester for conjugation to primary amines.
  • NOTA chelator for stable binding of radiometals.
  • Purity ≥97.0%.
  • CAS number 1338231-09-6.
  • Molecular formula C16H24N4O8; molecular weight 400.38 g/mol.
  • Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.