Learn More
Medchemexpress LLC NOTA-P2-RM26 | 1446005-10-2 | 97.2% | 1543.76 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP108961MG
Encompass_Preferred
NOTA-P2-RM26 is an antagonist of bombesin analogs. It targets the gastrin-releasing peptide receptor (GRPR/BB2), which is a molecular target for prostate cancer visualization. This product is utilized in imaging studies for prostate cancer using PET and SPECT/CT, where it is labeled with 111In and 68Ga.
  • Antagonist of bombesin analogs
  • Targets gastrin-releasing peptide receptor (GRPR/BB2)
  • Used in prostate cancer imaging studies
  • Can be labeled with 111In and 68Ga for PET and SPECT/CT applications
Catalog No. 50-004-24068