Learn More
Electron Microscopy Sciences Synaptek NOTCH Grid with 2x1 mm slot, Beryllium-Copper
SDP
Supplier:  Electron Microscopy Sciences S2010NOT
Encompass_Preferred
Synaptek unflexible grids are made of a special Beryllium-Copper alloy and very reliable under the electron beam. Offer extreme stability for coating with support film. 4 mil thick (100um), 3.05 mm diameter. Contamination free and reusable after cleaning.
I.D.: 2x1 mm slot
Material: Beryllium-Copper
Catalog No. 50-291-71