Learn More
Sino Biological NOTCH1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 50654T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse NOTCH1 (rM NOTCH1; Catalog#50654-M08B; NP_032740.3; Met1-Gln526). NOTCH1 specific IgG was purified by Mouse NOTCH1 affinity chromatography.
Catalog No. 50-224-8005