Learn More
AdipoGen Notch1 (mouse):Fc (human) (rec.) (3 x 50 µg)
SDP
Supplier:  AdipoGen AG40B01093050
Encompass_Preferred
Protein. The extracellular domain of mouse Notch1 (aa 19-488) (12 epidermal growth factor-like (EGF) repeats) is fused at the C-terminus to the Fc portion of human IgG1. Source: CHO cells. Endotoxin content: <0.1EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds to mouse DLL4. Purity: >95% (SDS-PAGE). Notch signaling pathway regulates many different cell fate decisions in both vertebrate and invertebrate species. There are 5 canonical Notch ligands in mammals: Jagged-1, Jagged-2, DLL1, DLL3 and DLL4. These can bind to the four Notch receptors Notch 1-4. It is important for pattern formation during development such as neurogenesis, angiogenesis or myogenesis and regulates T cell development and stem cell maintenance. Notch signaling is also involved in cellular processes through-out adulthood. Signaling via Notch occurs between neighbouring cells and both the receptor and its ligands are transmembrane proteins.
Catalog No. 50-205-3175