Learn More
Aviva Systems Biology Corp NOTCH2 Antibody - middle region (ARP32723_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32723_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NOTCH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NOTCH2. Alternate names: AGS2, hN2, HJCYSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54727