Learn More
AdipoGen anti-Notch2, mAb (16F11) (100 µg)
SDP
Supplier:  AdipoGen AG20B0052C100
Encompass_Preferred
Monoclonal Antibody. Recognizes human and mouse endogenous Notch-2 receptor. Isotype: Rat IgG1kappa. Clone: 16F11. Applications: FACS. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. Notch signaling pathway regulates many different cell fate decisions in both vertebrate and invertebrate species. There are 5 canonical Notch ligands in mammals: Jagged-1, Jagged-2, DLL1, DLL3 and DLL4. These can bind to the four Notch receptors Notch 1-4. It is important for pattern formation during development such as neurogenesis, angiogenesis or myogenesis and regulates T cell development and stem cell maintenance. Notch signaling is also involved in cellular processes through-out adulthood. Signaling via Notch occurs between neighbouring cells and both the receptor and its ligands are transmembrane proteins.
Catalog No. 50-205-2963